Product Details

SNP ID
rs7179261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42512022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGTCCATTAATATCAAAGAACAC[A/C]TTTGCTCCTTAGTGCCATTGCTGGT
Phenotype
MIM: 602534
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SNAP23 PubMed Links

Gene Details

Gene
SNAP23
Gene Name
synaptosome associated protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003825.3 Intron NP_003816.2
NM_130798.2 Intron NP_570710.1
XM_006720725.2 Intron XP_006720788.1
XM_017022692.1 Intron XP_016878181.1
XM_017022693.1 Intron XP_016878182.1
XM_017022694.1 Intron XP_016878183.1

View Full Product Details