Product Details

SNP ID
rs3218569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:626730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGATGGGCATGTTGGACCCTGAC[A/G]GGCAGAGCAGACCCGCGCTGAGCCT
Phenotype
MIM: 606839 MIM: 182099
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDHR5 PubMed Links

Gene Details

Gene
CDHR5
Gene Name
cadherin related family member 5
There are no transcripts associated with this gene.

Gene
SCT
Gene Name
secretin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021920.3 326 Missense Mutation NP_068739.1
XM_017018109.1 326 Missense Mutation XP_016873598.1

View Full Product Details