Product Details
- SNP ID
-
rs3218568
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:626920 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCCACCAGGCCCTGTAGCAGCCG[C/T]TGGAGCCGCGCGCCCTCCCGCAGGC
- Phenotype
-
MIM: 606839
MIM: 182099
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CDHR5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147980762] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDHR5
- Gene Name
- cadherin related family member 5
There are no transcripts associated with this gene.
- Gene
- SCT
- Gene Name
- secretin
View Full Product Details