Product Details
- SNP ID
-
rs4852914
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:73207437 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGATAGCCCAATGGCTCACTCCCT[C/T]ATCCCCATCAAGATTTGACTTAGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NOTO
PubMed Links
Gene Details
- Gene
- NOTO
- Gene Name
- notochord homeobox
- Gene
- SMYD5
- Gene Name
- SMYD family member 5
There are no transcripts associated with this gene.
View Full Product Details