Product Details

SNP ID
rs3211664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:89311205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGGACTGCGGTCGTTAGTCTCC[C/G]GCGAGTTGTTGCCTGGGCTGGACGT
Phenotype
MIM: 116901 MIM: 607693
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CKS2 PubMed Links

Gene Details

Gene
CKS2
Gene Name
CDC28 protein kinase regulatory subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001827.2 Intron NP_001818.1
Gene
MIR3153
Gene Name
microRNA 3153
There are no transcripts associated with this gene.

Gene
SECISBP2
Gene Name
SECIS binding protein 2
There are no transcripts associated with this gene.

View Full Product Details