Product Details

SNP ID
rs3793024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42932411 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCACAGAGGAAGGAGAACCTGCCC[A/G]GAAAAAAAGAGTGGGTGGATGTCAG
Phenotype
MIM: 610774 MIM: 606817
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNPY3 PubMed Links

Gene Details

Gene
CNPY3
Gene Name
canopy FGF signaling regulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318842.1 Intron NP_001305771.1
NM_001318845.1 Intron NP_001305774.1
NM_001318847.1 Intron NP_001305776.1
NM_001318848.1 Intron NP_001305777.1
NM_006586.4 Intron NP_006577.2
Gene
CNPY3-GNMT
Gene Name
CNPY3-GNMT readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318856.1 Intron NP_001305785.1
NM_001318857.1 Intron NP_001305786.1
NM_001318858.1 Intron NP_001305787.1
Gene
PTCRA
Gene Name
pre T-cell antigen receptor alpha
There are no transcripts associated with this gene.

View Full Product Details