Product Details

SNP ID
rs16967574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42207461 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATGGTAATTGTGTGGGTTTTCAC[A/G]CAAGAGAGATAACACACGCAGGTAT
Phenotype
MIM: 604260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STAT5B PubMed Links

Gene Details

Gene
STAT5B
Gene Name
signal transducer and activator of transcription 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012448.3 Intron NP_036580.2
XM_005257626.4 Intron XP_005257683.1
XM_017024977.1 Intron XP_016880466.1

View Full Product Details