Product Details

SNP ID
rs16969397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78004493 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACACTGAGGAGTCAAGACGCTAT[C/T]CTGTCAGCCAGGGGCTTTCCTAGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBC1D2B PubMed Links

Gene Details

Gene
TBC1D2B
Gene Name
TBC1 domain family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015079.5 Intron NP_055894.6
NM_144572.1 Intron NP_653173.1
XM_011521387.2 Intron XP_011519689.1
XM_017022022.1 Intron XP_016877511.1

View Full Product Details