Product Details

SNP ID
rs17037047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:158126020 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATTTCAGCAATAACAGACGTAAC[A/C]GTGAAAAGACAGGAGAGAATGTATT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM198B PubMed Links

Gene Details

Gene
FAM198B
Gene Name
family with sequence similarity 198 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031700.2 3378 UTR 3 NP_001026870.2
NM_001128424.1 3378 UTR 3 NP_001121896.1
NM_016613.6 3378 UTR 3 NP_057697.2

View Full Product Details