Product Details

SNP ID
rs16840060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:207823924 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAATCCAATGAAATTAGAATCGTC[C/T]ACCAATAAGGAGACTACTGGGTGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHM3 PubMed Links

Gene Details

Gene
PLEKHM3
Gene Name
pleckstrin homology domain containing M3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080475.2 7171 UTR 3 NP_001073944.1
XM_005246545.3 7171 UTR 3 XP_005246602.1
XM_006712521.3 7171 Intron XP_006712584.1
XM_011511160.2 7171 UTR 3 XP_011509462.1
XM_011511163.2 7171 Intron XP_011509465.1
XM_017004071.1 7171 Intron XP_016859560.1
XM_017004072.1 7171 UTR 3 XP_016859561.1
XM_017004073.1 7171 UTR 3 XP_016859562.1

View Full Product Details