Product Details

SNP ID
rs16839491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98264031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTCACCCTACATCCATGTCCCA[A/G]ACAGATTTTTTTCATTACATTCAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5H6 PubMed Links

Gene Details

Gene
OR5H6
Gene Name
olfactory receptor family 5 subfamily H member 6 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005479.1 Intron NP_001005479.1
XM_011513118.2 Intron XP_011511420.1

View Full Product Details