Product Details

SNP ID
rs17022333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:94269152 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAGAAGGTTGACTTGTAATCAAA[C/G]TTAAGTGTGAAATGGTTTTTTGAAC
Phenotype
MIM: 615847 MIM: 604259
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEP83 PubMed Links

Gene Details

Gene
CEP83
Gene Name
centrosomal protein 83
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042399.1 Intron NP_001035858.1
NM_016122.2 Intron NP_057206.2
XM_006719437.2 Intron XP_006719500.1
XM_011538424.1 Intron XP_011536726.1
XM_011538427.2 Intron XP_011536729.1
XM_011538429.2 Intron XP_011536731.1
XM_017019385.1 Intron XP_016874874.1
XM_017019386.1 Intron XP_016874875.1
XM_017019387.1 Intron XP_016874876.1
XM_017019388.1 Intron XP_016874877.1
XM_017019389.1 Intron XP_016874878.1
Gene
PLXNC1
Gene Name
plexin C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005761.2 Intron NP_005752.1
XM_006719186.3 Intron XP_006719249.1
XM_011537730.2 Intron XP_011536032.1
XM_011537731.2 Intron XP_011536033.1
XM_017018671.1 Intron XP_016874160.1
XM_017018672.1 Intron XP_016874161.1

View Full Product Details