Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_024776.3 | 7327 | UTR 3 | NP_079052.2 | ||
XM_005254670.2 | 7327 | UTR 3 | XP_005254727.1 | ||
XM_005254671.2 | 7327 | UTR 3 | XP_005254728.1 | ||
XM_005254673.2 | 7327 | UTR 3 | XP_005254730.1 | ||
XM_005254674.2 | 7327 | UTR 3 | XP_005254731.1 | ||
XM_005254675.4 | 7327 | UTR 3 | XP_005254732.1 | ||
XM_011522033.2 | 7327 | UTR 3 | XP_011520335.1 | ||
XM_011522034.2 | 7327 | UTR 3 | XP_011520336.1 | ||
XM_011522035.2 | 7327 | UTR 3 | XP_011520337.1 | ||
XM_011522036.2 | 7327 | UTR 3 | XP_011520338.1 | ||
XM_011522037.2 | 7327 | UTR 3 | XP_011520339.1 | ||
XM_011522038.2 | 7327 | UTR 3 | XP_011520340.1 | ||
XM_011522039.2 | 7327 | UTR 3 | XP_011520341.1 | ||
XM_011522040.1 | 7327 | Intron | XP_011520342.1 | ||
XM_011522041.1 | 7327 | Intron | XP_011520343.1 | ||
XM_011522042.2 | 7327 | UTR 3 | XP_011520344.1 |