Product Details

SNP ID
rs16953854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74551688 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAATCTTAAATTTTACCTGAGTCA[A/G]TTGTTAACAAGGAAATGAGAGACCA
Phenotype
MIM: 612457
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARID3B PubMed Links

Gene Details

Gene
ARID3B
Gene Name
AT-rich interaction domain 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307939.1 Intron NP_001294868.1
NM_006465.3 Intron NP_006456.1

View Full Product Details