Product Details

SNP ID
rs16978165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74675481 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTAGGGCATGCATCTTGGGCTCCA[A/C]AGCAGCTTCAATGGGTGTCACATTC
Phenotype
MIM: 609956
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RAB37 PubMed Links

Gene Details

Gene
RAB37
Gene Name
RAB37, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006638.2 Intron NP_001006639.1
NM_001163989.1 Intron NP_001157461.1
NM_001163990.1 Intron NP_001157462.1
NM_175738.4 Intron NP_783865.1
XM_005257288.4 Intron XP_005257345.1

View Full Product Details