Product Details

SNP ID
rs16855703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:136769788 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCCACCTACTAGAAAAGGTTAGA[C/T]ATTGACCTAGATGTGGAGATCTGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THSD7B PubMed Links

Gene Details

Gene
THSD7B
Gene Name
thrombospondin type 1 domain containing 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316349.1 Intron NP_001303278.1
XM_017005049.1 Intron XP_016860538.1

View Full Product Details