Product Details

SNP ID
rs12532096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75438689 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCACAGACCAGGAGCACTTTAGC[G/T]TTCCTGGTGGCCACATCACACTGCT
Phenotype
MIM: 615754
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POM121C PubMed Links

Gene Details

Gene
POM121C
Gene Name
POM121 transmembrane nucleoporin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099415.2 Intron NP_001092885.2

View Full Product Details