Product Details

SNP ID
rs17844245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140795310 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGATCGTGGACAGGCCGCTGCAG[G/T]TTTTCCATGTGGAAGTGGAGGTGAA
Phenotype
MIM: 606307 MIM: 606308 MIM: 606309
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCDHA1 PubMed Links

Gene Details

Gene
PCDHA1
Gene Name
protocadherin alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018900.3 452 Intron NP_061723.1
NM_031410.2 452 Intron NP_113598.1
NM_031411.2 452 Intron NP_113599.1
Gene
PCDHA2
Gene Name
protocadherin alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018905.2 452 Missense Mutation GTT,TTT V116F NP_061728.1
NM_031495.1 452 Missense Mutation GTT,TTT V116F NP_113683.1
NM_031496.1 452 Missense Mutation GTT,TTT V116F NP_113684.1
Gene
PCDHA3
Gene Name
protocadherin alpha 3
There are no transcripts associated with this gene.

View Full Product Details