Product Details

SNP ID
rs17854944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7090043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTTACGGAGGAGGAGGCCGCT[A/G]CGAGGAGTACCGGGGCTACTCACCC
Phenotype
MIM: 609665 MIM: 605444
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NLRP14 PubMed Links

Gene Details

Gene
NLRP14
Gene Name
NLR family pyrin domain containing 14
There are no transcripts associated with this gene.

Gene
RBMXL2
Gene Name
RNA binding motif protein, X-linked like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014469.4 1110 Missense Mutation TAC,TGC Y308C NP_055284.3

View Full Product Details