Product Details

SNP ID
rs17364024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:213285452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACATGTAACTCCTTTATTCAGGG[A/C]GGTAAGAAAGAGATGATTATCTGTT
Phenotype
MIM: 612173
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC100130451 PubMed Links

Gene Details

Gene
LOC100130451
Gene Name
uncharacterized LOC100130451
There are no transcripts associated with this gene.

Gene
SPAG16
Gene Name
sperm associated antigen 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025436.2 695 Intron NP_001020607.1
NM_024532.4 695 Intron NP_078808.3
XM_006712746.2 695 Intron XP_006712809.1
XM_011511814.1 695 Intron XP_011510116.1
XM_011511815.1 695 Intron XP_011510117.1
XM_011511816.2 695 Intron XP_011510118.1
XM_011511817.1 695 Intron XP_011510119.1
XM_011511818.1 695 Intron XP_011510120.1
XM_011511819.2 695 Intron XP_011510121.1
XM_011511820.1 695 Intron XP_011510122.1
XM_011511821.2 695 UTR 5 XP_011510123.1
XM_011511822.1 695 Intron XP_011510124.1
XM_011511823.2 695 Intron XP_011510125.1
XM_011511824.1 695 Intron XP_011510126.1
XM_011511827.1 695 Intron XP_011510129.1
XM_011511828.1 695 Intron XP_011510130.1
XM_011511829.1 695 Intron XP_011510131.1
XM_011511831.1 695 Intron XP_011510133.1
XM_011511832.1 695 Intron XP_011510134.1
XM_011511835.2 695 Intron XP_011510137.1
XM_011511837.2 695 Intron XP_011510139.1
XM_017004896.1 695 Intron XP_016860385.1
XM_017004897.1 695 Intron XP_016860386.1
XM_017004898.1 695 Intron XP_016860387.1
XM_017004899.1 695 Intron XP_016860388.1
XM_017004900.1 695 Intron XP_016860389.1
XM_017004901.1 695 Intron XP_016860390.1
XM_017004902.1 695 Intron XP_016860391.1

View Full Product Details