Product Details

SNP ID
rs35788500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55346423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGACCGCCGCCTTCCTGCTGAC[C/T]CTACTGCTGCAGCTCCTGCCGCCCG
Phenotype
MIM: 611715
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRD5A3 PubMed Links

Gene Details

Gene
SRD5A3
Gene Name
steroid 5 alpha-reductase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024592.4 203 Silent Mutation ACC,ACT T29T NP_078868.1
XM_005265766.3 203 Silent Mutation ACC,ACT T29T XP_005265823.1
XM_005265767.3 203 Silent Mutation ACC,ACT T29T XP_005265824.1
XM_017008601.1 203 Intron XP_016864090.1

View Full Product Details