Product Details

SNP ID
rs17354643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:83094012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTACAAAGAGTAGAAAAATTGT[A/G]TTCTTCCCATACTTAATATATATTC
Phenotype
MIM: 607515
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLAC8 PubMed Links

Gene Details

Gene
PLAC8
Gene Name
placenta specific 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130715.1 Intron NP_001124187.1
NM_001130716.1 Intron NP_001124188.1
NM_016619.2 Intron NP_057703.1

View Full Product Details