Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_182530.2 | Intron | NP_872336.2 | |||
XM_011539052.2 | Intron | XP_011537354.1 | |||
XM_011539055.1 | Intron | XP_011537357.1 | |||
XM_017018967.1 | Intron | XP_016874456.1 | |||
XM_017018968.1 | Intron | XP_016874457.1 | |||
XM_017018969.1 | Intron | XP_016874458.1 | |||
XM_017018970.1 | Intron | XP_016874459.1 | |||
XM_017018971.1 | Intron | XP_016874460.1 | |||
XM_017018972.1 | Intron | XP_016874461.1 | |||
XM_017018973.1 | Intron | XP_016874462.1 | |||
XM_017018974.1 | Intron | XP_016874463.1 | |||
XM_017018975.1 | Intron | XP_016874464.1 | |||
XM_017018976.1 | Intron | XP_016874465.1 |