Product Details

SNP ID
rs17114777
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:57000600 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAAATACATGTTGTAAGGCTAAA[C/T]ATGTAACTTTATTGTGTGTTTGTGT
Phenotype
MIM: 603448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAB1 PubMed Links

Gene Details

Gene
DAB1
Gene Name
DAB1, reelin adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021080.3 Intron NP_066566.3
Gene
LOC105378749
Gene Name
uncharacterized LOC105378749
There are no transcripts associated with this gene.

View Full Product Details