Product Details

SNP ID
rs17163868
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:26321890 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACCTGGCAGCATATTAGAAAATAG[C/T]TTATGTTACAACAAAAACCCTATAA
Phenotype
MIM: 114280 MIM: 609151
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIM1L PubMed Links

Gene Details

Gene
AIM1L
Gene Name
absent in melanoma 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039775.3 5200 UTR 3 NP_001034864.2
XM_005245918.2 5200 UTR 3 XP_005245975.1
XM_011541672.1 5200 UTR 3 XP_011539974.1
XM_011541673.2 5200 UTR 3 XP_011539975.1
Gene
CD52
Gene Name
CD52 molecule
There are no transcripts associated with this gene.

Gene
UBXN11
Gene Name
UBX domain protein 11
There are no transcripts associated with this gene.

View Full Product Details