Product Details

SNP ID
rs16979840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15354942 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACATGACAAGCTCATACCACTGAG[C/T]GTTTTGGTCCTACCACAATATATAT
Phenotype
MIM: 300091
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FIGF PubMed Links

Gene Details

Gene
FIGF
Gene Name
c-fos induced growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004469.4 Intron NP_004460.1
Gene
PIR-FIGF
Gene Name
PIR-FIGF readthrough
There are no transcripts associated with this gene.

View Full Product Details