Product Details

SNP ID
rs17004613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:43879965 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTGGGCCTCTCCCTATGCTCCTG[A/G]GGTGGAACTGAGAAGACGTGGGTTC
Phenotype
MIM: 614794
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGPAT3 PubMed Links

Gene Details

Gene
AGPAT3
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037553.1 Intron NP_001032642.1
NM_020132.4 Intron NP_064517.1
XM_005261160.4 Intron XP_005261217.1
XM_006724029.3 Intron XP_006724092.1
XM_006724030.3 Intron XP_006724093.1
XM_006724031.3 Intron XP_006724094.1
XM_011529660.2 Intron XP_011527962.1
XM_011529661.2 Intron XP_011527963.1
XM_011529662.2 Intron XP_011527964.1
XM_011529663.2 Intron XP_011527965.1
XM_011529664.2 Intron XP_011527966.1
XM_011529665.2 Intron XP_011527967.1
XM_017028408.1 Intron XP_016883897.1
XM_017028409.1 Intron XP_016883898.1
XM_017028410.1 Intron XP_016883899.1
XM_017028411.1 Intron XP_016883900.1
XM_017028412.1 Intron XP_016883901.1
XM_017028413.1 Intron XP_016883902.1
XM_017028414.1 Intron XP_016883903.1

View Full Product Details