Product Details

SNP ID
rs16872526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:75379892 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACTGTACCCAGCTGGACCAATTA[G/T]GATCTTTTAAGTCCCAACCCTTCAT
Phenotype
MIM: 604677
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COL4A3BP PubMed Links

Gene Details

Gene
COL4A3BP
Gene Name
collagen type IV alpha 3 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130105.1 Intron NP_001123577.1
NM_005713.2 Intron NP_005704.1
NM_031361.2 Intron NP_112729.1
XM_006714513.2 Intron XP_006714576.1
XM_011543090.2 Intron XP_011541392.1
XM_011543091.2 Intron XP_011541393.1
XM_017008919.1 Intron XP_016864408.1

View Full Product Details