Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001032372.1 | Intron | NP_001027544.1 | |||
NM_001032373.1 | Intron | NP_001027545.1 | |||
NM_001032374.1 | Intron | NP_001027546.1 | |||
NM_001146220.2 | Intron | NP_001139692.1 | |||
NM_001319088.1 | Intron | NP_001306017.1 | |||
NM_001319089.1 | Intron | NP_001306018.1 | |||
NM_001319090.1 | Intron | NP_001306019.1 | |||
NM_015919.3 | Intron | NP_057003.2 | |||
NM_016444.2 | Intron | NP_057528.2 | |||
XM_005259227.2 | Intron | XP_005259284.1 | |||
XM_006723367.3 | Intron | XP_006723430.1 | |||
XM_006723368.3 | Intron | XP_006723431.1 | |||
XM_006723369.2 | Intron | XP_006723432.1 | |||
XM_017027262.1 | Intron | XP_016882751.1 | |||
XM_017027263.1 | Intron | XP_016882752.1 | |||
XM_017027264.1 | Intron | XP_016882753.1 | |||
XM_017027265.1 | Intron | XP_016882754.1 |