Product Details

SNP ID
rs16988877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51077997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCTGACGAGGTCCATCCCACC[A/G]TGAAGACCATCATTTGTCCCGCATC
Phenotype
MIM: 606135
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLK14 PubMed Links

Gene Details

Gene
KLK14
Gene Name
kallikrein related peptidase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311182.1 998 UTR 3 NP_001298111.1
NM_022046.5 998 UTR 3 NP_071329.2
XM_017026820.1 998 Intron XP_016882309.1

View Full Product Details