Product Details

SNP ID
rs16834245
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:152622929 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTTGGAGCTCTGGCAACGGCAAC[A/G]GTGAGACCTGCGGCATCTGTGGTGA
Phenotype
MIM: 612613 MIM: 612614
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCE3A PubMed Links

Gene Details

Gene
LCE3A
Gene Name
late cornified envelope 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178431.1 175 Missense Mutation CGT,TGT R59C NP_848518.1
Gene
LCE3B
Gene Name
late cornified envelope 3B
There are no transcripts associated with this gene.

View Full Product Details