Product Details
- SNP ID
-
rs6609714
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:48347476 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAATAGCACATCTGCAGGGATGCAG[A/G]GGATGAGTGGAAGGTTCTCTTACGG
- Phenotype
-
MIM: 300325
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SSX3
PubMed Links
Gene Details
- Gene
- SSX3
- Gene Name
- SSX family member 3
View Full Product Details