Product Details

SNP ID
rs17075508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:173020759 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAGTCAGCCATCCGGACAGAACC[A/G]TTCCTGTGATGGTGTTACACTGCTG
Phenotype
MIM: 603931 MIM: 611331
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0E1 PubMed Links

Gene Details

Gene
ATP6V0E1
Gene Name
ATPase H+ transporting V0 subunit e1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003945.3 Intron NP_003936.1
Gene
SNORA74B
Gene Name
small nucleolar RNA, H/ACA box 74B
There are no transcripts associated with this gene.

View Full Product Details