Product Details

SNP ID
rs17108103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148269618 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGTCCAAGATTGTACAGGACAGC[C/T]CTCAGTTTACACCTGTTACCTCAGA
Phenotype
MIM: 615205
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102546294 PubMed Links

Gene Details

Gene
LOC102546294
Gene Name
uncharacterized LOC102546294
There are no transcripts associated with this gene.

Gene
SPINK13
Gene Name
serine peptidase inhibitor, Kazal type 13 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040129.2 Intron NP_001035218.1

View Full Product Details