Product Details

SNP ID
rs16965650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44666934 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTATACTGCCTCCTAGAAGAAAC[C/T]GAAACTCTAGGGCACTATAGAAAAA
Phenotype
MIM: 614661 MIM: 610844
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PATL2 PubMed Links

Gene Details

Gene
PATL2
Gene Name
PAT1 homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145112.1 Intron NP_001138584.1
XM_011521336.2 Intron XP_011519638.2
XM_011521337.2 Intron XP_011519639.2
XM_011521338.2 Intron XP_011519640.1
XM_011521339.2 Intron XP_011519641.1
XM_011521340.2 Intron XP_011519642.1
XM_011521341.1 Intron XP_011519643.1
XM_011521342.2 Intron XP_011519644.1
XM_011521343.2 Intron XP_011519645.1
XM_011521344.2 Intron XP_011519646.1
XM_011521345.2 Intron XP_011519647.1
XM_011521346.2 Intron XP_011519648.2
XM_011521347.1 Intron XP_011519649.1
XM_011521348.2 Intron XP_011519650.1
XM_017022000.1 Intron XP_016877489.1
XM_017022001.1 Intron XP_016877490.1
Gene
SPG11
Gene Name
spastic paraplegia 11 (autosomal recessive)
There are no transcripts associated with this gene.

View Full Product Details