Product Details

SNP ID
rs16828757
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:23847115 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAACTCAATACCAATGAAGGTGTGC[A/G]GGAAGGAGGCAGTCTATAAAACTCT
Phenotype
MIM: 612280
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FUCA1 PubMed Links

Gene Details

Gene
FUCA1
Gene Name
fucosidase, alpha-L- 1, tissue
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000147.4 Intron NP_000138.2
XM_005245821.2 Intron XP_005245878.1
XM_011541167.2 Intron XP_011539469.1
XM_017000905.1 Intron XP_016856394.1

View Full Product Details