Product Details

SNP ID
rs17101109
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:77979784 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTCTAAAAGTATGACTGCCCTTT[C/T]CTTTATCCAAAAACGTGATTGTTCT
Phenotype
MIM: 611327 MIM: 603444
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJB4 PubMed Links

Gene Details

Gene
DNAJB4
Gene Name
DnaJ heat shock protein family (Hsp40) member B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317099.1 Intron NP_001304028.1
NM_001317100.1 Intron NP_001304029.1
NM_001317101.1 Intron NP_001304030.1
NM_001317102.1 Intron NP_001304031.1
NM_001317103.1 Intron NP_001304032.1
NM_007034.4 Intron NP_008965.2
Gene
FUBP1
Gene Name
far upstream element binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303433.1 Intron NP_001290362.1
NM_003902.4 Intron NP_003893.2
XM_011542391.2 Intron XP_011540693.1
XM_011542392.2 Intron XP_011540694.1
XM_011542393.2 Intron XP_011540695.1
XM_017002739.1 Intron XP_016858228.1
XM_017002740.1 Intron XP_016858229.1
XM_017002741.1 Intron XP_016858230.1
XM_017002742.1 Intron XP_016858231.1
XM_017002743.1 Intron XP_016858232.1
XM_017002744.1 Intron XP_016858233.1

View Full Product Details