Product Details

SNP ID
rs17357621
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:46342025 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCATGTACCCTTGGAAACTGGAC[C/T]CATGATTTCGCCTCCTCCTACACAT
Phenotype
MIM: 615394
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NSUN4 PubMed Links

Gene Details

Gene
NSUN4
Gene Name
NOP2/Sun RNA methyltransferase family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256127.1 Intron NP_001243056.1
NM_001256128.1 Intron NP_001243057.1
NM_199044.3 Intron NP_950245.2

View Full Product Details