Product Details

SNP ID
rs17059312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:28708601 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAAATGATGTTTTTTTGGGTTGT[G/T]AGTACTTAAAAACAAAACAATCCTC
Phenotype
MIM: 605744
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EXTL3 PubMed Links

Gene Details

Gene
EXTL3
Gene Name
exostosin like glycosyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001440.3 Intron NP_001431.1
XM_011544440.2 Intron XP_011542742.1
Gene
EXTL3-AS1
Gene Name
EXTL3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details