Product Details

SNP ID
rs34089965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46382705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTCAGTGCCAGGAGACCATCCG[C/G]GTCACCAAGCCCTGCACCCCCAAGA
Phenotype
MIM: 118495 MIM: 601441 MIM: 162096
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHRM4 PubMed Links

Gene Details

Gene
CHRM4
Gene Name
cholinergic receptor muscarinic 4
There are no transcripts associated with this gene.

Gene
DGKZ
Gene Name
diacylglycerol kinase zeta
There are no transcripts associated with this gene.

Gene
MDK
Gene Name
midkine (neurite growth-promoting factor 2)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012333.2 751 Silent Mutation CGC,CGG R121R NP_001012333.1
NM_001012334.2 751 Silent Mutation CGC,CGG R121R NP_001012334.1
NM_001270550.1 751 Silent Mutation CGC,CGG R121R NP_001257479.1
NM_001270551.1 751 Silent Mutation CGC,CGG R121R NP_001257480.1
NM_001270552.1 751 Silent Mutation CGC,CGG R65R NP_001257481.1
NM_002391.4 751 Silent Mutation CGC,CGG R121R NP_002382.1
XM_011520116.2 751 Silent Mutation CGC,CGG R121R XP_011518418.1
XM_017017764.1 751 Silent Mutation CGC,CGG R121R XP_016873253.1
Gene
MIR4688
Gene Name
microRNA 4688
There are no transcripts associated with this gene.

View Full Product Details