Product Details
- SNP ID
-
rs17726944
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:50947468 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTTCTGGGTTCAGATACTGAGAAC[A/G]TTTGATGCTTGGGAATTGATGGTCT
- Phenotype
-
MIM: 603503
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADNP-AS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116747043] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADNP-AS1
- Gene Name
- ADNP antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- DPM1
- Gene Name
- dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit
View Full Product Details