Product Details

SNP ID
rs17149957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87843538 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAACAAATCCAAAAGTTTCTCAG[A/C]AAAATATGTTTTTGTAACATAAAAA
Phenotype
MIM: 610821
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A40 PubMed Links

Gene Details

Gene
SLC25A40
Gene Name
solute carrier family 25 member 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018843.3 Intron NP_061331.2

View Full Product Details