Product Details

SNP ID
rs12720360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10352903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCAAGTGACCCCAGCACCCCCT[G/C]AGACTGCACCCCGCCTGGTCACCAG
Phenotype
MIM: 176941
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
TYK2 PubMed Links

Gene Details

Gene
TYK2
Gene Name
tyrosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003331.4 Intron NP_003322.3
XM_011528245.1 Intron XP_011526547.1
XM_011528246.2 Intron XP_011526548.1
XM_011528247.1 Intron XP_011526549.1
XM_011528249.2 Intron XP_011526551.1
XM_011528251.1 Intron XP_011526553.1
XM_011528252.1 Intron XP_011526554.1

View Full Product Details