Product Details

SNP ID
rs17549110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10372891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGTGGGAGAGGCAGCCTTGGG[A/G]ATATCTGAATTGCCTTTAGGATCTC
Phenotype
MIM: 611817
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLRC4-KLRK1 PubMed Links

Gene Details

Gene
KLRC4-KLRK1
Gene Name
KLRC4-KLRK1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199805.1 1051 UTR 3 NP_001186734.1
Gene
KLRK1
Gene Name
killer cell lectin like receptor K1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007360.3 1051 UTR 3 NP_031386.2
Gene
LOC101928100
Gene Name
uncharacterized LOC101928100
There are no transcripts associated with this gene.

View Full Product Details