Product Details

SNP ID
rs17026998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:97468472 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCATCCTTACTAATATCTCTAAGC[C/T]GTTCATTCATGCCCTTTATGTTAGT
Phenotype
MIM: 607045
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RMST PubMed Links

Gene Details

Gene
RMST
Gene Name
rhabdomyosarcoma 2 associated transcript (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details