Product Details

SNP ID
rs16974673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84983913 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGCAGCAGCTTGCTCTTGTGCT[A/T]TCGCAGTGCCGTTAGGTCCCTGAGT
Phenotype
MIM: 602972 MIM: 606207
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PDE8A PubMed Links

Gene Details

Gene
PDE8A
Gene Name
phosphodiesterase 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243137.1 Intron NP_001230066.1
NM_002605.2 Intron NP_002596.1
NM_173454.1 Intron NP_775656.1
XM_011521668.2 Intron XP_011519970.1
XM_017022308.1 Intron XP_016877797.1
XM_017022309.1 Intron XP_016877798.1
XM_017022310.1 Intron XP_016877799.1
XM_017022311.1 Intron XP_016877800.1
Gene
SLC28A1
Gene Name
solute carrier family 28 member 1
There are no transcripts associated with this gene.

View Full Product Details