Product Details

SNP ID
rs17147016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68929508 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACACCAGGAATAATCATATTGCA[A/T]GTCACCCTATCACTTAAGGTTATAG
Phenotype
MIM: 616382
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
UGT2A3 PubMed Links

Gene Details

Gene
UGT2A3
Gene Name
UDP glucuronosyltransferase family 2 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024743.3 1920 UTR 3 NP_079019.3
XM_011532247.2 1920 UTR 3 XP_011530549.1

View Full Product Details