Product Details
- SNP ID
-
rs16989893
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:32122034 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAGGAGGAGGAGGCACTTGCCAC[A/G]CACGCTGGAGACTCGAGCCTGCCTT
- Phenotype
-
MIM: 182380
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AP1B1P1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138991067] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AP1B1P1
- Gene Name
- adaptor related protein complex 1 beta 1 subunit pseudogene 1
There are no transcripts associated with this gene.
- Gene
- SLC5A1
- Gene Name
- solute carrier family 5 member 1
There are no transcripts associated with this gene.
View Full Product Details