Product Details

SNP ID
rs17847440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7669377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTGTCCTGGGGCTGTTGGTGTC[A/T]AGCAAGACCCTGTGCTCCATGGAAG
Phenotype
MIM: 609565 MIM: 605565
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MCEMP1 PubMed Links

Gene Details

Gene
MCEMP1
Gene Name
mast cell expressed membrane protein 1
There are no transcripts associated with this gene.

Gene
RETN
Gene Name
resistin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193374.1 88 Silent Mutation TCA,TCT S17S NP_001180303.1
NM_020415.3 88 Silent Mutation TCA,TCT S17S NP_065148.1

View Full Product Details