Product Details

SNP ID
rs17238318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:55540368 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTTTTGGTTGGGAAATATCAGC[A/G]CAAGTTAAAAGTTACTATATAATTT
Phenotype
MIM: 606902
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107984745 PubMed Links

Gene Details

Gene
LOC107984745
Gene Name
uncharacterized mitochondrial protein AtMg00860-like
There are no transcripts associated with this gene.

Gene
PYGO1
Gene Name
pygopus family PHD finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015617.2 7310 UTR 3 NP_056432.1
XM_005254300.4 7310 UTR 3 XP_005254357.1
XM_011521446.2 7310 UTR 3 XP_011519748.1

View Full Product Details